Learn More
Socorex™ Bottle Top Dispenser 520 Model
Brand: Socorex™ 525.100
606.50 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Specifications
Bottletop Dispenser | |
EN ISO 8655 | |
Ground Glass plunger |
Autoclavable | |
10 to 100 mL | |
525 |