Learn More
Lenz Laborglasinstrumente™ Borosilicate Glass Separating Funnels with PTFE Plug
Made of DURAN™ borosilicate glass. Lenz Laborglasinstrumente™ Borosilicate Glass Separating Funnels with PTFE Plug are used in liquid-liquid extractions to separate the components of a mixture into immiscible solvent phases of different densities.
124.00€ - 400.00€
Product Code | Brand | Capacity (Metric) | Joint Number | Pore Size | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Joint Number | Pore Size | Price | Quantity & Availability | |||
10102051
|
Lenz Laborglasinstrumente™
04030170 |
1000 mL | 29/32 NS | 6mm |
400.00€
Pack of 2 |
Estimated Shipment: 26-09-2025
Log in to see stock. |
|||
10373371
|
Lenz Laborglasinstrumente™
04026137 |
100 mL | 29/32 NS | 2.5mm |
124.00€
Pack of 2 |
Estimated Shipment: 24-09-2025
Log in to see stock. |
|||
10731191
|
Lenz Laborglasinstrumente™
04030137 |
100 mL | 19/26 NS | 2.5mm |
229.00€
Pack of 2 |
In Stock
Log in to see stock. ![]() |
|||
12840453
|
Lenz Laborglasinstrumente™
04030149 |
250 mL | 29/32 NS | 4mm |
209.00€
Pack of 2 |
In Stock
Log in to see stock. ![]() |
|||
10694192
|
Lenz Laborglasinstrumente™
04030158 |
500 mL | 29/32 NS | 4mm |
317.00€
Pack of 2 |
Estimated Shipment: 19-09-2025
Log in to see stock. |
|||
10461711
|
Lenz Laborglasinstrumente™
04030128 |
50 mL | 19/26 NS | 2.5mm |
229.00€
Pack of 2 |
Estimated Shipment: 24-09-2025
Log in to see stock. |
|||
Description
- With NS socket and PE stopper
- Without scale, with PTFE plug
- With screwthread retaining device
- With inscription label
Specifications
Borosilicate Glass |