Learn More
Hamilton™ Terasaki Dispenser
This multi-syringe dispenser accurately dispenses six samples simultaneously up to 50 times with the press of a lever.
Brand: Hamilton™ 201900
976.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Designed for rapid filling of 60 well HLA/Terasaki plates with 6.35mm well spacing
- Uses removable needles
- Available for a range of volumes
Specifications
1/Ea. | |
Liquid or gas sampling | |
PTFE (Plunger) |
Gastight Syringe | |
Model 1702 Terasaki Dispenser | |
25 μL |