Learn More
Brand™ Borosilicate Glass Needle Valve Stopcock
Suitable for Novus desiccators with lid socket. BRAND™ Borosilicate Glass Needle Valve Stopcock employs a lateral venting hole which helps to reduce turbulence.
Brand:  Brand™ 82415
47.40 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10176540
  • 53.05€ / Each
Only null left
Estimated Shipment: 04-09-2025
Description
  • Suitable for Novus desiccators with lid socket.
  • Needle-valve stopcock PTFE, 0-4 mm.
  • Lateral venting hole helps to reduce turbulence.
  • Nozzle with 10mm outer Ø.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 Each
Borosilicate Glass
With Lid Socket
Needle Valve Stopcock
For Desiccators