Learn More
VACUUBRAND™ Borosilicate Glass Flask With Spherical Ground Joint KS25/25
Brand:  VACUUBRAND™ 20638497
71.58 EUR valid until 2025-08-24
Use promo code "24140" to get your promotional price.
Product Code. 10218571
  • 75.35€ / Each
Quality Analysis and Quality Control Testing
Use promo code 24140 at checkout
View Available Promotions
Only null left
Estimated Shipment: 21-07-2025
Specifications
500 mL
Borosilicate Glass
1 Each
Flask