Learn More
Lenz Laborglasinstrumente™ Separating Funnels with Glass Plug, without Scale
For use in liquid-liquid extractions to separate components. Lenz Laborglasinstrumente™ Separating Funnels, with Glass Plug, without Scale are manufactured from durable DURAN™ borosilicate glass.
79.00€ - 158.00€
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
15330895
|
Lenz Laborglasinstrumente™
04000049 |
250 mL |
90.00€
Pack of 2 |
Estimated Shipment: 24-09-2025
Log in to see stock. |
|||||
10728731
|
Lenz Laborglasinstrumente™
04000037 |
100 mL |
90.00€
Pack of 2 |
Estimated Shipment: 24-09-2025
Log in to see stock. |
|||||
10465601
|
Lenz Laborglasinstrumente™
04000070 |
1000 mL |
158.00€
Pack of 2 |
Estimated Shipment: 22-09-2025
Log in to see stock. |
|||||
10475601
|
Lenz Laborglasinstrumente™
04000073 |
2000 mL |
124.00€
Each |
Estimated Shipment: 24-09-2025
Log in to see stock. |
|||||
15330885
|
Lenz Laborglasinstrumente™
04000058 |
500 mL |
102.00€
Pack of 2 |
Estimated Shipment: 24-09-2025
Log in to see stock. |
|||||
10507551
|
Lenz Laborglasinstrumente™
04000028 |
50 mL |
79.00€
Pack of 2 |
Estimated Shipment: 08-10-2025
Log in to see stock. |
|||||
Description
- Without scale; with solid glass plug
- With NS socket and PE stopper
- With screwthread retaining device
- Corrosion-free and resistant to thermal shock
Specifications
Borosilicate Glass | |
Polyethylene |