Learn More
Lenz Laborglasinstrumente™ Separating Funnels, with PTFE Plug and Scale
For use in liquid-liquid extractions to separate components. Lenz Laborglasinstrumente™ Separating Funnels, with PTFE Plug and Scale are manufactured from durable DURAN™ borosilicate glass.
81.00€ - 190.00€
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10643161
|
Lenz Laborglasinstrumente™
04006137 |
100 mL |
88.00€
Pack of 2 |
Estimated Shipment: 22-09-2025
Log in to see stock. |
|||||
11852672
|
Lenz Laborglasinstrumente™
04006170 |
1000 mL |
171.00€
Pack of 2 |
Estimated Shipment: 17-09-2025
Log in to see stock. |
|||||
11836084
|
Lenz Laborglasinstrumente™
04006173 |
2000 mL |
190.00€
Each |
Estimated Shipment: 16-09-2025
Log in to see stock. |
|||||
10100121
|
Lenz Laborglasinstrumente™
04006149 |
250 mL |
107.00€
Pack of 2 |
Estimated Shipment: 15-09-2025
Log in to see stock. |
|||||
10393511
|
Lenz Laborglasinstrumente™
04006128 |
50 mL |
81.00€
Pack of 2 |
Estimated Shipment: 16-09-2025
Log in to see stock. |
|||||
11871593
|
Lenz Laborglasinstrumente™
04006158 |
500 mL |
130.00€
Pack of 2 |
Estimated Shipment: 16-09-2025
Log in to see stock. |
|||||
Description
- With scale (approx. volume/mL)
- With PTFE plug
- With NS socket and PE stopper
- With screwthread retaining device
- Corrosion-free and resistant to thermal shock
Specifications
Borosilicate Glass | |
Polyethylene |