Learn More
Grant Instruments™ Test Tube Rack for Unstirred Water Baths
Keeps test tubes upright and secure in the bath.
Brand: Grant Instruments™ J2-13
142.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Keeps test tubes upright and secure in the bath.
- Compatible with a wide range of Grant Unstirred Water Baths
- Holds up to 105 test tubes depending on size
- Dual slotted trays keep tubes secure and prevent tipping
Specifications
55 13 mm Tubes |