Learn More
Agilent BioTek 50 TS Waste Bottle
Provides waste containment
Brand:  Agilent BioTek 48140
59.74 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 11583495
  • 64.40€ / Each
Description
  • Capacity: 2L
  • Without cap
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2 L
Waste Bottle