Learn More
Agilent BioTek Accessories For MultiFlo FX Microplate Dispenser RAD peri cassette
GREENER_CHOICE
Accurate and precise peristaltic pump dispensing.
Brand:  Agilent BioTek 1260017
465.56 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 16592092
  • 502.00€ / Each
Only null left
Estimated Shipment: 25-09-2025
Description
For use with MultiFlo FX Microplate Dispenser RAD peri cassette
TRUSTED_SUSTAINABILITY
Specifications
Specifications
500 nL to 50 μL
MultiFlo FX Microplate Dispenser
1 μL
Plastic