Learn More
Mölnlycke™ Biogel™ NeoTech Gloves
Sterile cleanroom gloves with beaded cuffs and curved fingers are suitable for use in aseptic and Class 100 (ISO 5) / EU GMP Grade A environments. Adviced to be worn in combination with the Biogel™ NeoTech Indicator™ Underglove to form a breach indication system.
396.00€ - 495.00€
Specifications
Certifications/Compliance | ASTM D3577 Viral penetration ASTM F1671 Pyrogenicity ASTM D7102 |
---|---|
Certifications/Compliance (Europe) | EN 455, ISO 10282 , EN 374, EN 16523-1, PPE Cat. III, Quality/Environment ISO 13485, ISO 14001, Sterilisation ISO 11137, Viral penetration ISO 16604, Allergenicity ISO 10993, Labelling EN 556-1, EN ISO 15223-1, EN ISO 21420 |
Cleanroom Class | Class 100 (ISO 5) |
Accelerator Free | No |
AQL | 0.65 |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17895728
![]() |
Mölnlycke™
4450955-02 |
5.5 |
495.00€
200 pairs |
Estimated Shipment: 11-09-2025
Log in to see stock. |
|||||
17885728
![]() |
Mölnlycke™
4450960-02 |
6 |
495.00€
200 pairs |
Estimated Shipment: 11-09-2025
Log in to see stock. |
|||||
17865778
![]() |
Mölnlycke™
4450965-02 |
6.5 |
495.00€
200 pairs |
Estimated Shipment: 11-09-2025
Log in to see stock. |
|||||
17855738
![]() |
Mölnlycke™
4450970-02 |
7 |
495.00€
200 pairs |
Estimated Shipment: 11-09-2025
Log in to see stock. |
|||||
17895778
![]() |
Mölnlycke™
4450975-02 |
7.5 |
495.00€
200 pairs |
Estimated Shipment: 11-09-2025
Log in to see stock. |
|||||
17805738
![]() |
Mölnlycke™
4450980-02 |
8 |
495.00€
200 pairs |
Estimated Shipment: 11-09-2025
Log in to see stock. |
|||||
17875738
![]() |
Mölnlycke™
4450985-02 |
8.5 |
495.00€
200 pairs |
Estimated Shipment: 11-09-2025
Log in to see stock. |
|||||
17885778
![]() |
Mölnlycke™
4450990-02 |
9 |
396.00€
160 pairs |
Estimated Shipment: 11-09-2025
Log in to see stock. |
|||||
Description
- Can be worn together with Biogel™ NeoTech Indicator™ Underglove to form a breach indication system
- Tested and cleared for use with chemotherapy agents
- Synthetic polychloroprene
- Biogel hydrogel polymer coating for easy donning
- Exceed industry standard for force at break, tensile strength and elongation
- Every glove (100%) is air-inflation tested for holes typically not detected in a visual inspection
Specifications
ASTM D3577 Viral penetration ASTM F1671 Pyrogenicity ASTM D7102 | |
Class 100 (ISO 5) | |
0.65 | |
Single-use | |
8.3 mil | |
Specific | |
Polychloroprene | |
0.19 mm | |
Sterile | |
0.17 mm | |
11.8 in. |
EN 455, ISO 10282 , EN 374, EN 16523-1, PPE Cat. III, Quality/Environment ISO 13485, ISO 14001, Sterilisation ISO 11137, Viral penetration ISO 16604, Allergenicity ISO 10993, Labelling EN 556-1, EN ISO 15223-1, EN ISO 21420 | |
No | |
White | |
Smooth | |
0.21 mm | |
Yes | |
7.5 mil | |
Powder Free | |
6.7 mil (Cuff) | |
21 MPa | |
30 cm |