Learn More
Lenz Laborglasintrumente™ Bent Socket Adapter
Adapters with NS socket and bent glass olive (90°)
Brand: Lenz Laborglasintrumente™ 05103003
12.88 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- 1 per pack
- Made of DURAN™ tubing
Specifications
NS 24/29 | |
Adapter with NS socket and bent olive | |
Duran Tubing | |
1 |