Learn More
Bel-Art™ SP Scienceware™ Glove Box Clamping Rings
Adjustable rings hold gloves in place
30.40€ - 36.10€
Specifications
Product Type | Clamping Ring |
---|---|
For Use With (Application) | Fasten Glove Firmly to Glove Box |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
11775084
|
Bel-Art™
H50029-0200 |
Fits 6 in. Circular Port Holes | N/A | ||||||
11702156
|
Bel-Art™
H50025-0320 |
Fits 8 in. Circular Port Holes |
36.10€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Fit Techni-Dome™ glove boxes(Mfr. No. 500300000) or other glove boxes with circular port holes of the correct diameter
Specifications
Clamping Ring | |
Fasten Glove Firmly to Glove Box |