Learn More
BD BBL™ Dehydrated Culture Media: PPLO Agar (Mycoplasma Agar)
Click to view available options
:
PPLO Agar (Use with Mycoplasma Supplement); 500g
Specifications
Specifications
Content And Storage | <30°C |
Form | Dehydrated |
Media Type | PPLO Agar |
Product Type | Agar |
Formulation | Beef heart infusion from 50g 6g/L, Peptone 10g/L, Sodium Chloride 5g/L, Agar 14g/L |
For Use With (Application) | Used with Mycoplasma supplement or Mycoplasma supplement S for isolation and cultivation of Mycoplasma |
Packaging Type | Bottle |
Quantity | 500 g |