Learn More
BD BBL™ Dehydrated Culture Media: Brucella Broth
Click to view available options
:
Brucella Broth; 500g
Description
- Dehydrated
- Culture Media
- Brucella Broth
Specifications
Specifications
Certifications/Compliance | CCAM, ISO, USDA |
Form | Dehydrated |
Media Type | Brucella Broth |
Product Type | Microbiology Media |
Formulation | Pancreatic digest of Casein 10g/L, Peptic digest of Animal Tissue 10g/L, Dextrose 1g/L, Yeast extract 2g/L, Sodium Chloride 5g/L, Sodium Bisulfite 0.1g/L |
For Use With (Application) | Cultivation of Brucella organisms |
Packaging Type | Bottle |
Quantity | 500 g |