Learn More
Thermo Scientific™ Barnstead™ E-Pure™ Cartridge Kits
Replace worn, outdated and otherwise ineffective cartridges with Thermo Scientific™ Barnstead™ E-Pure™ Cartridge Kits.
Brand:  Thermo Scientific™ D50223
 View more versions of this product
Product Code. 11796289
  • 176.00€ / Pack of 3
The Academic Best Buy Book 2025
Use promo code 25081 at checkout
View Available Promotions
Only null left
Estimated Shipment: 10-09-2025
Description
Replace worn, outdated and otherwise ineffective cartridges with Thermo Scientific™ Barnstead™ E-Pure™ Cartridge Kits. These cartridge kits are classified based on type of feed water used; choose the appropriate kit according to your feed water and need for organic free water.
For the Nanopure, E-pure Water Systems
TRUSTED_SUSTAINABILITY
Specifications
Specifications
E-Pure Ultrapure Water Purification System, Cleaning Cartridge
Cleaning Cartridge