Learn More
Azlon™ PTFE Sleeves
Fabricated from pure PTFE for superior chemical, corrosion and contamination resistance, high thermal stability and good flexural properties. Azlon™ PTFE Sleeves form a grease-free gas tight seal for ground glass joints. Available in standard joint sizes for accurate fit.
19.20€ - 46.80€
Specifications
Type | Truncated cone |
---|---|
For Use With (Application) | For low vacuum work |
Material | PTFE |
Product Type | Sleeves |
Temperature Range (Metric) | –100 to 250°C |
Product Code | Brand | Standard Taper Joint No. | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint No. | Price | Quantity & Availability | |||||
11363714
|
Azlon
SWN961 |
14/23 |
19.30€
Pack of 10 |
Estimated Shipment: 05-11-2025
Log in to see stock. |
|||||
11373714
|
Azlon
SWN962 |
19/26 |
19.20€
Pack of 10 |
Estimated Shipment: 15-10-2025
Log in to see stock. |
|||||
11383714
|
Azlon
SWN963 |
24/29 |
22.60€
Pack of 10 |
Estimated Shipment: 05-11-2025
Log in to see stock. |
|||||
10223610
|
Azlon
SWN965 |
34/35 |
24.30€
Pack of 10 |
Estimated Shipment: 05-11-2025
Log in to see stock. |
|||||
12613072
|
Azlon
SWN966 |
40/38 |
37.75€
Pack of 10 |
Estimated Shipment: 05-11-2025
Log in to see stock. |
|||||
12541456
|
Azlon
SWN967 |
45/50 |
46.80€
Pack of 10 |
Estimated Shipment: 05-11-2025
Log in to see stock. |
|||||
11393714
|
Azlon
SWN964 |
29/32 |
21.55€
Pack of 10 |
Estimated Shipment: 18-11-2025
Log in to see stock. |
|||||
10446860
|
Azlon
SWN969 |
55/44 |
44.55€
Pack of 10 |
Estimated Shipment: 05-11-2025
Log in to see stock. |
|||||
Description
Fabricated from pure PTFE for superior chemical, corrosion and contamination resistance, high thermal stability and good flexural properties. Azlon™ PTFE Sleeves form a grease-free gas tight seal for ground glass joints. Available in standard joint sizes for accurate fit.
- Truncated cone of pure PTFE
- Allows easy separation of joints
- Excellent chemical resistance
- Outstanding resistance to fatigue
- Working temperature range of -100°C to +250°C
- No contamination from joint grease
- Intended for single or occasional use
- Appropriate for low vacuum work
- USP Class VI grade PTFE
- Verified for food contact applications per US regulations
Specifications
Truncated cone | |
PTFE | |
–100 to 250°C |
For low vacuum work | |
Sleeves |