Learn More
Axygen™ Single Well Medium Profile Reagent Reservoirs
Manufactured with virgin polypropylene for excellent chemical resistance
360.09€ - 430.28€
Specifications
Certifications/Compliance | ANSI/SLAS, RNase/DNase-free, non-pyrogenic |
---|---|
Color | Clear |
Capacity (Metric) | 170 mL |
Material | Polypropylene |
Type | Reagent Reservoir |
Product Code | Brand | Sterility | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sterility | Price | Quantity & Availability | |||||
12527877
![]() |
Axygen™
RES-SW384-FX |
Non-sterile |
360.09€
Pack of 25 |
Estimated Shipment: 14-11-2025
Log in to see stock. |
|||||
12537877
![]() |
Axygen™
RES-SW384-FX-SI |
Sterile |
430.28€
Pack of 25 |
Estimated Shipment: 14-11-2025
Log in to see stock. |
|||||
Description
Corning offers a complete line of Axygen single-well and multiple-well reagent reservoirs which are available in multiple formats for automated and manual use. All reservoirs are designed to comply with standard ANSI/SLAS footprint dimensions for microplates and are compatible with most automation workstations. Reagent reservoirs are manufactured with polypropylene for excellent chemical resistance.
- Durable and cost effective
- Various formats for multiple applications
Specifications
ANSI/SLAS, RNase/DNase-free, non-pyrogenic | |
170 mL | |
Reagent Reservoir | |
Single-use |
Clear | |
Polypropylene | |
384 |