Learn More
Trajan Scientific and Medical™ Autosampler Syringes with Fixed Needle for Thermo Scientific GCs
Click to view available options
Volume (Metric):
5 μL
10 μL
Needle Point Style:
Bevel
Cone
Product Type:
Fixed Needle
Removable Needle
Unit Size:
Each
Description
- High quality solutions for liquid handling
- Accurate and reproducible injections
- Specifically designed for Thermo Scientific instruments
Specifications
Specifications
Description | CTC/Thermo (Classic Button) Syringe |
Needle Gauge | 26 G |
Diameter (Metric) Outer Needle | 0.47 mm |
Product Type | Fixed Needle |
Volume (Metric) | 10 μL |
Needle Point Style | Cone |
For Use With (Equipment) | Thermo Gas Chromatography |