Learn More
Cole-Parmer™ PVDF Compression to Male Threaded Adapter Fittings
These compression fittings are an excellent choice to connect rigid tubing systems, with higher pressure resistance than that provided by barbed fittings.
Brand: Cole-Parmer™ 06385-85
This item is not returnable.
View return policy
Description
- For rigid tubing systems, including tubing made from fluoropolymers, other rigid plastics, and metals
- Assemble the fitting to tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal
- No leaks, flow restrictions and costly downtime
- Sterilization by autoclave or ethylene oxide (ETO)
Specifications
Autoclavable | |
3/8 tubing O.D. x 3/8 in. NPT(M) Thread | |
11/32 in. orifice, 1-13/32 x 13/16 in. | |
3/8 in. NPT(M) |
PVDF | |
1 Each | |
Autoclave or Ethylene Oxide | |
Compression Fitting |