Learn More
Cole-Parmer™ Parker Hannifin™ PFA Union Connector Compression Fitting
Compression fittings are an excellent choice to connect rigid tubing systems, able to withstand higher pressures than barbed fittings.
Brand:  Cole-Parmer™ 31321-64
103.54 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 11730379
  • 113.00€ / Each
Only null left
Estimated Shipment: 02-10-2025
Description
  • Available in straight, "T", and elbow shapes for unmatched versatility
  • Provided with a nut that compresses rigid tubing against the fitting body for high-pressure applications
  • Excellent for use with tubing manufactured from fluoropolymers, other rigid plastics and metals
  • Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal
  • Sterilization through autoclaving or ethylene oxide (ETO)
Specifications
Specifications
Autoclavable
6mm tubing O.D.
Autoclave or Ethylene Oxide
Compression Fitting
PFA
1 Each
Straight