Learn More
SymLine Continuous Level Control Accessories
Accessories for Continuous Level Control
Brand: SymLine 108099
165.36 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Measuring filling levels continuously - Monitoring non-transparent canisters on a permanent basis - No problem with the continuous level control unit!Specifications
Antistatic floor mat, ethylenevinylacetate, dimensions (W x H x D): 610 x 1220 x 3 mm | |
610 mm | |
Eliminating/Dissipating Static Charge |
1.22 m | |
Ground cable | |
Antistatic Floor Mat |