Learn More
Buerkle™ Anti-Static Set for Decanting Flammables
Buerkle™ Anti-Static Set for Decanting Flamables ensures easy and reliable earthing.
Brand: Buerkle™ 5602-1000
56.95 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- When decanting flammable liquids without the machines and containers involved in the process being sufficiently earthed, electrostatic build-up can occur, which for many substances can result in fire or an explosion.
- Anti-static set ensures easy and reliable earthing
- The Anti-static set consists of three colour-coded connecting copper cables with clamps in a handy plastic case
Specifications
Electric Conductive Pumps | |
For easy and reliable earthing | |
(3) Copper Cable; Clamp |