Learn More
Amberlite™ IRC-748 ion-exchange resin, Thermo Scientific Chemicals
Product Code. 10153821
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic Bottle
CAS: 79620-28-3
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 79620-28-3
MDL Number MFCD00132702
Specifications
Physical Form Beads
Particle Size Particle size analysis: uniformity coefficient: <1.70,% <0.300mm: <1.0%,%>1.000mm: <5.0%, harmonic mean size: 0.500 - 0.650mm
Packaging Plastic Bottle
Water Retention Capacity (Na+): 60 to 65%
Density 0.7500g/mL
Merck Index 14, 382
Quantity 1 kg
Chemical Name or Material Amberlite™ IRC-748
Product Identifier
  • Amberlite]TS IRC-748 - ion-exchange resin