Learn More
Aluminum hexafluoro-2,4-pentanedionate, 98+%, Thermo Scientific™
Product Code. p-7058825
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
In MOCVD
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 15306-18-0
Molecular Formula C15H3AlF18O6
Molecular Weight (g/mol) 648.14
MDL Number MFCD00050051
InChI Key SINRSQHVMMWHSI-UHFFFAOYSA-N
Synonym aluminum hexafluoroacetylacetonate, aluminum hexafluoropentanedionate, aluminum hexafluoroacetylacetonate, 3z-4-bis 2z-1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-yl oxy alumanyl oxy-1,1,1,5,5,5-hexafluoropent-3-en-2-one
IUPAC Name aluminium(3+) tris(1,1,1,5,5,5-hexafluoro-2,4-dioxopentan-3-ide)
SMILES [Al+3].FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F.FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F.FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F
Specifications
Boiling Point 170°C (decomposition)
Melting Point 70°C to 73°C
Physical Form Crystalline
Quantity 2 g
Formula Weight 648.17
Percent Purity ≥98%
Sensitivity Moisture sensitive
Chemical Name or Material Aluminum hexafluoro-2,4-pentanedionate
compliance-icons
Product Identifier
  • Aluminum hexafluoro-2,4-pentanedionate
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .