Learn More
Cole Parmer™ Aluminum Crimpled-Walled Weighing Dishes with Tab
Crimpled walls for rigidity and a side finger tab handle for easy carrying.
23.40€ - 26.25€
Specifications
Bottom Shape | Flat |
---|---|
Disposable | Single-use |
Material | Aluminum |
Shape | Round |
Type | Weighing Dish |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
15406083
|
Cole Parmer™
01018-51 |
20 mL |
26.25€
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15496073
|
Cole Parmer™
01018-50 |
60 mL |
23.40€
Pack of 100 |
Estimated Shipment: 01-08-2025
Log in to see stock. |
|||||
15416083
|
Cole Parmer™
01018-52 |
80 mL |
23.85€
Pack of 100 |
Estimated Shipment: 01-08-2025
Log in to see stock. |
|||||
Description
- Disposable.
- Crimpled wall for rigidity.
- Finger tab handle for improved handling, labeling.
- Sturdy gauge aluminum for heavy duty use.
- Angled sides for easy separation.
- No oil residue, no vinyl coating to create contamination problems.
- Smooth bottom.
- May be used in a wide variety of applications including weighing, moisture testing, wastewater analysis, chemical, and pharmaceutical uses.
Specifications
Flat | |
Aluminum | |
Weighing Dish | |
100/Pk. |
Single-use | |
Round | |
Tab |