Learn More
Al-23 Tube, Both Ends Open, O.D.: 7mm, I.D.: 4mm, Thermo Scientific Chemicals
35.07€ - 125.63€
Specifications
Chemical Name or Material Al-23 Tube
Name Note Both ends open
View More Specs
Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11333937
View Documents
Thermo Scientific Alfa Aesar
031558.DM
100 mm
35.07€
100mm
11393937
View Documents
Thermo Scientific Alfa Aesar
031558.DR
200 mm This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A
11373937
View Documents
Thermo Scientific Alfa Aesar
031558.DW
300 mm
60.40€
300mm
11383937
View Documents
Thermo Scientific Alfa Aesar
031558.EH
600 mm This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A
11343937
View Documents
Thermo Scientific Alfa Aesar
031558.EM
800 mm This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A
11353937
View Documents
Thermo Scientific Alfa Aesar
031558.EO
1000 mm
125.63€
1000mm
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Al-23 Tube
Both ends open
Videos
Safety and Handling
Safety and Handling
missing translation for 'tsca' : U
Recommended Storage : Ambient temperatures