Learn More
Eppendorf™ Aerosol-tight Caps for Round Buckets, S-4x400 Rotor
Shop All Eppendorf Products
Click to view available options
For Use With (Equipment):
Round Buckets, S-4x400 Rotor
Unit Size:
Pack of 2
Description
- Suitable for round buckets used in S-4x400 Rotor
- Provides containment of hazardous materials
Specifications
Specifications
For Use With (Equipment) | Round Buckets, S-4x400 Rotor |
Includes | 2 Caps |