Learn More
Eppendorf™ Adapter for Conical Tubes, Bottles, and Plates
Conical tube, bottle, and plate adapter for rotor S-4xUniversal-Large.
Brand: Eppendorf™ 5920755005
440.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- 2 pieces
- Adapter
Specifications
Adapter | |
50 mL conical tubes, 175 to 250 mL bottles, and plates | |
For Rotor S-4xUniversal-Large | |
2 |