Learn More
3M™ VHB™ Acrylic Gray Foam Tape
Offering design flexibility with its viscoelasticity and powerful ability to bond to a variety of surfaces. 3M™ VHB™ Acrylic Gray Foam Tape creates a permanent seal against water, moisture and more.
Brand: 3M™ 7000088220
This item is not returnable.
View return policy
Description
- Virtually invisible fastening
- Can replace mechanical fasteners (rivets, welds, screws) or liquid adhesives
- Pressure sensitive adhesive bonds on contact to provide immediate handling strength
- Allows the use of thinner, lighter weight and dissimilar materials
Specifications
Acrylic | |
Acrylic | |
Up to 93°C | |
0.023 mm | |
33 m |
Gray | |
Foam tape | |
91 mil | |
33 m x 2.54 cm | |
2.54 cm |