Learn More
Cytiva AcroPak™ 400 Capsules with Fluorodyne™ II Membrane
Click to view available options
Pore Size:
0.2 μm
Unit Size:
Each
Description
- Double layer sterilizing membrane enhances reliability
- Upstream air vent prevents vapor lock
- Manufactured without the use of adhesives to minimize extractables
- Pre-sterilized by gamma irradiation
Specifications
Specifications
Material (Membrane) | Hydrophilic PVDF |
Pore Size | 0.2 μm |
Sterility | Sterile |
Filtration Area | 400 cm2 |
No. per Pack | 1/Ea. |
Outlet Style | Stepped Hose Barb |