Learn More
Agilent BioTek Accessories for Lionheart FX Automated Live Cell Imager
Miscellaneous accessories for use with Lionheart FX Automated Live Cell Imager.
Brand: Agilent BioTek 1450009
This item is not returnable.
View return policy
Description
Injector accessories with offset tips, for use with Lionheart FX. Comes with tip mount, tubing, priming trough and tubing bracket.
Specifications
Environment Control Cover | |
Lionheart FX Automated Live Cell Imager |