Learn More
Thermo Scientific™ Accessories for Cimarec+™ Stirrer Series
Get precise stirring control for your routine protocols.
20.50€ - 66.13€
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
15483660
|
Thermo Scientific™
CIC0001448 |
PT1000 External probe 0-400C |
66.13€
Each |
In Stock
Log in to see stock. |
|||||
15413660
|
Thermo Scientific™
CIC0001444 |
Transparent shield for 4x4 |
23.82€
Each |
Estimated Shipment: 14-11-2025
Log in to see stock. |
|||||
15443660
|
Thermo Scientific™
CIC0001445 |
Transparent shield for 7x7 |
20.50€
Each |
In Stock
Log in to see stock. |
|||||
15453660
|
Thermo Scientific™
CIC0001446 |
Transparent shield for 10x10 |
25.33€
Each |
In Stock
Log in to see stock. |
|||||
15626245
|
Thermo Scientific™
CIC0001830 |
Supporting Rod |
31.66€
Each |
In Stock
Log in to see stock. |
|||||
15636245
|
Thermo Scientific™
CIC0001831 |
Clamp and holder |
37.89€
Each |
In Stock
Log in to see stock. |
|||||
Description
These accessories are for use with the Cimarec+™ series digital stirrers. Our new Thermo Scientific™ Cimarec+™ series digital stirrers offer precise stirring control for your routine protocols. The Cimarec+ Series are available in three sizes, providing flexibility from microscale chemistries to production operation.
Advanced technology
- Stir-Trac features innovative engineering for slow-speed stirring, consistent speed control, and strong magnetic coupling
Flexible, robust design
- Top plate choices in ceramic or aluminum
- Easy-to-read LED display for stirring
- Raised display design protects electronics from spills
- Optional splash guard protection shield
- Variable sizes for different sample volumes
Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
LED |