Learn More
Integra Biosciences™ 8-channel Adapter
Click to view available options
Type:
8-Channel adapter with ejector for grip tips
8-Channel stainless steel adapter, 40mm
Unit Size:
Each
Description
- 8-Channel stainless steel aspiration adapter
- Flame sterilizable for maximal safety
- Fit a variety of tubes, flasks, bottles etc.
- Accommodate 12- to 96-well plates
- With a precisely dosed volume
- Without loss of drops
- Without risk of contamination
- Without causing tiredness
- All parts in contact with fluids are autoclavable
Specifications
Specifications
Material | Stainless Steel |
Type | 8-Channel stainless steel adapter, 40mm |
For Use With (Equipment) | VACUBOY Aspiration Tools |