missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-1-indanone, 97+%

Product Code. p-7028410
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 34598-49-7 | C9H7BrO | 211.058 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34598-49-7
Molecular Formula C9H7BrO
Molecular Weight (g/mol) 211.058
MDL Number MFCD00082718
InChI Key KSONICAHAPRCMV-UHFFFAOYSA-N
Synonym 5-bromo-1-indanone, 5-bromoindanone, 5-bromoindan-1-one, 5-bromo-2,3-dihydro-1h-inden-1-one, 1h-inden-1-one, 5-bromo-2,3-dihydro, 1-indanone, 5-bromo, 5-bromo-indanone, 5-bromo indanone, zlchem 204, 5-bromo-1-oxoindane
PubChem CID 520695
IUPAC Name 5-bromo-2,3-dihydroinden-1-one
SMILES C1CC(=O)C2=C1C=C(C=C2)Br

Specifications

Melting Point 125°C to 129°C
Quantity 1 g
Beilstein 2357199
Formula Weight 211.06
Percent Purity 98%
Chemical Name or Material 5-Bromo-1-indanone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.