Learn More
4-Carboxybenzaldehyde, 96%, Thermo Scientific Chemicals
26.23€ - 824.00€
Chemical Identifiers
CAS | 619-66-9 |
---|---|
Molecular Formula | C8H6O3 |
Molecular Weight (g/mol) | 150.13 |
MDL Number | MFCD00006951 |
InChI Key | GOUHYARYYWKXHS-UHFFFAOYSA-N |
Synonym | 4-carboxybenzaldehyde, benzoic acid, 4-formyl, terephthalaldehydic acid, p-formylbenzoic acid, p-carboxybenzaldehyde, terephthaldehydic acid, 4-carboxybenzaladehyde, 4-formyl-benzoic acid, 4-formylbenzoicacid, unii-ues4qrk36e |
PubChem CID | 12088 |
IUPAC Name | 4-formylbenzoic acid |
SMILES | C1=CC(=CC=C1C=O)C(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10245580
|
Thermo Scientific Acros
154580050 |
5 g | Glass bottle |
26.23€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10497930
|
Thermo Scientific Acros
154580250 |
25 g | Glass bottle |
77.69€
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10786561
|
Thermo Scientific Acros
154581000 |
100 g | Glass bottle |
221.10€
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10004103
|
Thermo Scientific Acros
154585000 |
500 g | Glass bottle | N/A | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
619-66-9 | |
150.13 | |
GOUHYARYYWKXHS-UHFFFAOYSA-N | |
12088 | |
C1=CC(=CC=C1C=O)C(=O)O |
C8H6O3 | |
MFCD00006951 | |
4-carboxybenzaldehyde, benzoic acid, 4-formyl, terephthalaldehydic acid, p-formylbenzoic acid, p-carboxybenzaldehyde, terephthaldehydic acid, 4-carboxybenzaladehyde, 4-formyl-benzoic acid, 4-formylbenzoicacid, unii-ues4qrk36e | |
4-formylbenzoic acid |
Specifications
619-66-9 | |
100.0 | |
Yellow | |
95% min. (HPLC) | |
HO2CC6H4CHO | |
10, 671 | |
Solubility in water: soluble | |
C1=CC(=CC=C1C=O)C(=O)O | |
150.13 | |
150.13 | |
Fine Crystalline Powder |
95.0 | |
247°C | |
Authentic | |
C8H6O3 | |
MFCD00006951 | |
4-carboxybenzaldehyde, benzoic acid, 4-formyl, terephthalaldehydic acid, p-formylbenzoic acid, p-carboxybenzaldehyde, terephthaldehydic acid, 4-carboxybenzaladehyde, 4-formyl-benzoic acid, 4-formylbenzoicacid, unii-ues4qrk36e | |
GOUHYARYYWKXHS-UHFFFAOYSA-N | |
4-formylbenzoic acid | |
12088 | |
96% | |
4-Carboxybenzaldehyde |
Safety and Handling
GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
May cause damage to organs through prolonged or repeated exposure.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 210-607-4
- 4-Carboxybenzaldehyde
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H373-May cause damage to organs.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.