Learn More
4-Bromo-N-(4-methoxybenzyl)aniline, 97%, Thermo Scientific Chemicals
Product Code. 15442098
Click to view available options
Quantity:
250 mg
1 g
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 175357-73-0
Molecular Formula C14H14BrNO
Molecular Weight (g/mol) 292.176
MDL Number MFCD00017822
InChI Key LJKLYFDMLGYRCA-UHFFFAOYSA-N
Synonym 4-bromo-n-4-methoxyphenyl methyl aniline, 4-bromo-n-4-methoxybenzyl aniline, n-4-methoxybenzyl-4-bromoaniline, 4-bromo-phenyl-4-methoxy-benzyl-amine, 4-bromophenyl 4-methoxyphenyl methyl amine
PubChem CID 775227
IUPAC Name 4-bromo-N-[(4-methoxyphenyl)methyl]aniline
SMILES COC1=CC=C(C=C1)CNC2=CC=C(C=C2)Br
Specifications
Quantity 250 mg
Formula Weight 292.18
Percent Purity 97%
Chemical Name or Material 4-Bromo-N-(4-methoxybenzyl)aniline