Learn More
3M™ PAPR Battery Charger Cradle
Click to view available options
:
Battery charger cradle
Description
- Replacement part for the TR-341 and TR-344 battery charger kits
- 3M Catalog# TR-340
Specifications
Specifications
Product Type | Single-Unit Charging Station |
For Use With (Application) | Provides easy-to-use interface between all TR-300 batteries and all TR-900 series power supplies |
For Use With (Equipment) | Versaflo™ TR-300 and Speedglas™ TR-300 PAPR Systems |
Model | TR-340 |
No. of Ports | One |
Quantity | 1 Case |