Learn More
BRAND™ 384-Well Skirted Microplates, BIO-CERT™ PCR QUALITY
Optimized for reliable and precise results in a wide range of PCR applications.
Brand: BRAND™ 781358
374.80 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
BRAND™ PCR products deliver excellent, reproducible results for a wide range of PCR and qPCR applications. Manufactured from high-quality virgin resins, these products are free from lubricants, chemical additives, DNA, DNase, RNase, and other PCR inhibitors. Key features include:
- Exceptional Quality and Purity
- BIO-CERT™ PCR QUALITY ensures plates are free of human DNA, RNase, DNase, pyrogens, and PCR inhibitors
- Free of contaminants that could interfere with reagents and samples
- Ultra-smooth vessel interiors minimize enzyme nucleic acid binding
- Precision Thermal Performance
- Thin, uniform walls ensure consistent, precise thermal transfer for reliable results
- Designed for rapid and accurate heat transfer, reducing cycle times
- Wide Compatibility
- Suitable for most commercially available thermal cyclers
- White PCR plates, enhanced with titanium dioxide (TiO2), optimize fluorescence signal reflection and are recommended for qPCR applications
- User-Friendly Features
- Blue alphanumeric coding for easy sample identification
- Well rims designed to prevent cross-contamination and ensure reliable sealing with tailored mats
- Skirted plates provide extra rigidity, ideal for automated pipetting systems and transport
Specifications
No Barcode | |
Standard | |
16 x 24 | |
Bagged | |
Single-use | |
Thermal Cyclers | |
Polypropylene | |
384 | |
Non-treated | |
Conical Bottom (V-well) | |
Yes |
Untreated | |
Without Lid | |
Autoclavable | |
White | |
qPCR 384-well plate | |
Fully skirted | |
0.03 mL | |
Non-sterile | |
0.03 mL | |
Optimize results when performing quantitative RT-PCR |