Learn More
Greiner Bio-One 384-well Polystyrene Streptavidin-coated Flat Bottom Microplate
GREENER_CHOICE
Pre-blocked and ready-to-use.
1820.00€ - 4521.13€
Specifications
Format Standard
Array 16 x 24
Autoclavable Not Autoclavable
Material Polystyrene
No. of Wells 384
View More Specs
Products 5
Product Code Brand Color Price Quantity & Availability  
Product Code Brand Color Price Quantity & Availability  
10779214
GREENER_CHOICE
View Documents
Greiner Bio-One
781990
Clear
4276.57€
Pack of 40
Only null left
Estimated Shipment: 11-07-2025
Add to basket
11985315
GREENER_CHOICE
View Documents
Greiner Bio-One
781995
White This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A
13077943
GREENER_CHOICE
View Documents
Greiner Bio-One
781995
White
4364.44€
Pack of 40
Only null left
Estimated Shipment: 03-11-2025
Add to basket
10477092
GREENER_CHOICE
View Documents
Greiner Bio-One
781996
Black, Clear Bottom
4521.13€
Pack of 40
Only null left
Estimated Shipment: 03-11-2025
Add to basket
10554991
GREENER_CHOICE
View Documents
Greiner Bio-One
781997
Black
4521.13€
Pack of 40
Only null left
Estimated Shipment: 03-11-2025
Add to basket
Description
Description
  • Manufactured under DIN ISO 9001 guidelines
  • Biotin binding capacity: >1.5ng/well or >6pmol/well
  • Solid bottom
  • Storable at room temperature
  • With lid
Specifications
Specifications
Standard
Not Autoclavable
384
Streptavidin
Room Temperature
16 x 24
Polystyrene
Non-sterile
5
Videos
Safety and Handling
Safety and Handling
missing translation for 'shelfLife' : 3 Years after month of production