Learn More
RETSCH 360° Stainless Steel Ring Sieve For SR200/SR300 Rotor Beater Mills
Shop All Retsch Products
Click to view available options
Pore Size:
80 μm
1 mm
120 μm
1.5 mm
2 mm
200 μm
250 μm
3 mm
4 mm
5 mm
500 μm
6 mm
750 μm
8 mm
10 mm
Unit Size:
Each
Specifications
Specifications
Quantity | 1Each |
Pore Size | 80 μm |
Type | Ring Sieve |
Material | Stainless Steel |
For Use With (Equipment) | SR200/SR300 Rotor Beater Mills |