Learn More
Cole-Parmer™ 304 Stainless Steel Cover
For use with stainless steel beakers
36.70€ - 59.00€
Specifications
Material | Stainless Steel (304) |
---|---|
Quantity | 1Each |
Certifications/Compliance | AISI 304 |
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | |||||
11797974
|
Cole-Parmer™
07207-80 |
222mm |
59.00€
Each |
Estimated Shipment: 21-10-2025
Log in to see stock. |
|||||
11747974
|
Cole-Parmer™
07207-10 |
240mm |
40.55€
Each |
Estimated Shipment: 21-10-2025
Log in to see stock. |
|||||
11757974
|
Cole-Parmer™
07207-20 |
280mm |
36.70€
Each |
Estimated Shipment: 21-10-2025
Log in to see stock. |
|||||
11767974
|
Cole-Parmer™
07207-30 |
171.45 mm |
46.60€
Each |
Estimated Shipment: 21-10-2025
Log in to see stock. |
|||||
Description
- Covers reduce evaporation
- Contain harmful vapors, or keep contaminants out
Specifications
Stainless Steel (304) | |
1Each | |
AISI 304 |