missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-5-hydroxybenzeneboronic acid, 98%, Thermo Scientific™

Product Code. 15439377
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Fluoro-5-hydroxybenzeneboronic acid is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 871329-82-7
Molecular Formula C6H6BFO3
Molecular Weight (g/mol) 155.919
MDL Number MFCD07363778
InChI Key RMBFBZIEXCTPDB-UHFFFAOYSA-N
Synonym 3-fluoro-5-hydroxyphenyl boronic acid, 3-fluoro-5-hydroxybenzeneboronic acid, 3-fluoro-5-hydroxy-phenyl boronic acid, 3-fluoro-5-hydroxphenyl boronic acid, 3-hydroxyl-5-fluoro-phenyl-boronic acid, acmc-209qgp, 3-fluoro-5-hydroxyphenyl-boronic acid, 3-fluoranyl-5-oxidanyl-phenyl boronic acid, boronic acid,b-3-fluoro-5-hydroxyphenyl
PubChem CID 44717308
IUPAC Name (3-fluoro-5-hydroxyphenyl)boronic acid
SMILES B(C1=CC(=CC(=C1)F)O)(O)O

Specifications

Melting Point 54°C to 62°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 155.92
Percent Purity 98%
Chemical Name or Material 3-Fluoro-5-hydroxybenzeneboronic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.