missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethoxyphenol, 98%, Thermo Scientific Chemicals

Product Code. 11415956
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

3,5-dimethoxyphenol is a marker for poisoning from Taxus baccata.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-dimethoxyphenol is a marker for poisoning from Taxus baccata.

Solubility
Soluble in methanol, ethanol, benzene, water (slightly), and chloroform.

Notes
Air and light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 500-99-2
Molecular Formula C8H10O3
Molecular Weight (g/mol) 154.17
MDL Number MFCD00008388
InChI Key XQDNFAMOIPNVES-UHFFFAOYSA-N
Synonym taxicatigenin, phenol, 3,5-dimethoxy, phloroglucinol dimethyl ether, 1-hydroxy-3,5-dimethoxybenzene, 3,5-dimethoxy phenol, unii-23uxw8136a, phenol,5-dimethoxy, 3,5-dimethoxy-phenol, pubchem16421, acmc-209kk2
PubChem CID 10383
IUPAC Name 3,5-dimethoxyphenol
SMILES COC1=CC(OC)=CC(O)=C1

Specifications

Melting Point 45°C to 47°C
Boiling Point 172°C to 175°C (17 mmHg)
Flash Point 78°C (173°F)
Quantity 250 g
Beilstein 1239156
Sensitivity Air and light sensitive
Solubility Information Soluble in methanol,ethanol,benzene,water (slightly),and chloroform.
Formula Weight 154.16
Percent Purity 98%
Chemical Name or Material 3,5-Dimethoxyphenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.