Learn More
Tenak™ 22 mm Cryobox Dividers
Dividers for use with 32 mm high cryoboxes.
46.20€ - 55.80€
Specifications
Type | Grid Divider |
---|---|
For Use With (Equipment) | 32 mm high cryoboxes |
Height (Metric) | 22 mm |
Product Code | Brand | Holds | Tube Array | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Tube Array | Price | Quantity & Availability | ||||
15200624
|
Tenak™
TE22675 |
Vials with 15.50 mm O.D. | 8 x 8 cells |
51.00€
Pack of 36 |
Estimated Shipment: 19-09-2025
Log in to see stock. |
||||
15210624
|
Tenak™
TE22676 |
Vials with 17.75 mm O.D. | 7 x 7 cells |
51.00€
Pack of 36 |
Estimated Shipment: 19-09-2025
Log in to see stock. |
||||
15220534
|
Tenak™
TE22092 |
Vials with 13 mm O.D. | 9 x 9 cells |
46.20€
Pack of 36 |
Estimated Shipment: 19-09-2025
Log in to see stock. |
||||
15230534
|
Tenak™
TE22094 |
Vials with 10 mm O.D. | 12 x 12 cells |
51.00€
Pack of 36 |
Estimated Shipment: 19-09-2025
Log in to see stock. |
||||
15240534
|
Tenak™
TE22093 |
Vials with 12 mm O.D. | 10 x 10 cells |
46.20€
Pack of 36 |
Estimated Shipment: 17-07-2025
Log in to see stock. |
||||
15240614
|
Tenak™
TE22669 |
Vials with 7.50 mm O.D. | 16 x 16 cells |
55.80€
Pack of 36 |
Estimated Shipment: 19-09-2025
Log in to see stock. |
||||
15250534
|
Tenak™
TE22095 |
Vials with 9 mm O.D. | 13 x 13 cells |
51.00€
Pack of 36 |
Estimated Shipment: 19-09-2025
Log in to see stock. |
||||
15250614
|
Tenak™
TE22670 |
Vials with 8.75 mm O.D. | 14 x 14 cells |
55.80€
Pack of 36 |
Estimated Shipment: 19-09-2025
Log in to see stock. |
||||
15260614
|
Tenak™
TE22671 |
Vials with 9.50 mm O.D. | 13 x 13 cells |
55.80€
Pack of 36 |
Estimated Shipment: 19-09-2025
Log in to see stock. |
||||
15270614
|
Tenak™
TE22672 |
Vials with 10.25 mm O.D. | 12 x 12 cells |
55.80€
Pack of 36 |
Estimated Shipment: 19-09-2025
Log in to see stock. |
||||
11581665
|
Tenak™
TE22096 |
Vials with 8 mm O.D. | 14 x 14 cells |
51.00€
Pack of 36 |
Estimated Shipment: 19-09-2025
Log in to see stock. |
Hai già un account? Accedi per visualizzare il prezzo e la disponibilità. Sei nuovo da queste parti? Registrati qui! |
|||
11591665
|
Tenak™
TE22097 |
Vials with 7 mm O.D. | 16 x 16 cells |
51.00€
Pack of 36 |
Estimated Shipment: 19-09-2025
Log in to see stock. |
Hai già un account? Accedi per visualizzare il prezzo e la disponibilità. Sei nuovo da queste parti? Registrati qui! |
|||
15200534
|
Tenak™
TE22090 |
Vials with 17 mm O.D. | 7 x 7 cells |
46.20€
Pack of 36 |
Estimated Shipment: 19-09-2025
Log in to see stock. |
Hai già un account? Accedi per visualizzare il prezzo e la disponibilità. Sei nuovo da queste parti? Registrati qui! |
|||
Descrizione
- For use in cardboard cryoboxes
- Holds cryogenic vials
- Grid style divider
Specifica
Grid Divider | |
32 mm high cryoboxes | |
22 mm |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.