Learn More
RETSCH 203.2 dia. x 25.4mmH Stainless Steel Test Sieve, ASTM Certificate, Micrometer Pore Sizes
Shop All Retsch Products
Click to view available options
Pore Size:
20 μm
25 μm
32 μm
38 μm
45 μm
53 μm
63 μm
75 μm
90 μm
106 μm
125 μm
150 μm
180 μm
212 μm
250 μm
300 μm
355 μm
425 μm
500 μm
600 μm
710 μm
850 μm
Unit Size:
Each
Specifications
Specifications
Quantity | 1Each |
Pore Size | 106 μm |
Type | Test Sieve |
Certifications/Compliance | ASTM E 11 |
Material | Stainless Steel |
For Use With (Equipment) | Shaker AS 300 |
Height (Metric) | 25.4mm |
For Use With (Application) | For separation, fractioning, particle size determination |
Diameter (Metric) Outer | 203.2 mm |
Frame Material | Stainless Steel |