Learn More
RETSCH 200 dia. x 25mmH Stainless Steel Test Sieve
Shop All Retsch Products
Click to view available options
Pore Size:
1 mm
1.18 mm
1.4 mm
1.7 mm
2 mm
2.36 mm
2.8 mm
3.35 mm
355 μm
4 mm
425 μm
4.75 mm
500 μm
5.6 mm
600 μm
6.3 mm
6.7 mm
710 μm
8 mm
850 μm
9.5 mm
11.2 mm
12.5 mm
13.2 mm
16 mm
19 mm
22.4 mm
25 mm
26.5 mm
31.5 mm
37.5 mm
45 mm
50 mm
53 mm
63 mm
75 mm
90 mm
100 mm
106 mm
125 mm
Unit Size:
Each
Specifications
Specifications
Quantity | 1Each |
Pore Size | 1.18 mm |
Type | Test Sieve |
Certifications/Compliance | ASTM E 11 |
Material | Stainless Steel |
For Use With (Equipment) | Shaker AS 300 |
Height (Metric) | 25 mm |
For Use With (Application) | For separation, fractioning, particle size determination |
Diameter (Metric) Outer | 200 mm |
Frame Material | Stainless Steel |