missing translation for 'onlineSavingsMsg'
Learn More

2-Phenyl-2-pentanol, 98%, Thermo Scientific Chemicals

Product Code. 11445806
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4383-18-0
Molecular Formula C11H16O
Molecular Weight (g/mol) 164.248
MDL Number MFCD00046592
InChI Key YBQOTTAEVBHNEJ-UHFFFAOYSA-N
Synonym 2-phenyl-2-pentanol, 2-phenyl-pentan-2-ol, benzenemethanol, .alpha.-methyl-.alpha.-propyl, methyl phenylbutanol, acmc-1aei8, methyl phenyl n-propyl carbinol
PubChem CID 138214
IUPAC Name 2-phenylpentan-2-ol
SMILES CCCC(C)(C1=CC=CC=C1)O

Specifications

Quantity 25 g
Formula Weight 164.25
Percent Purity 98%
Chemical Name or Material 2-Phenyl-2-pentanol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.