Learn More
2-Phenyl-2-pentanol, 98%, Thermo Scientific Chemicals
Product Code. 11435806
Click to view available options
Quantity:
1 g
5 g
25 g
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4383-18-0
Molecular Formula C11H16O
Molecular Weight (g/mol) 164.248
MDL Number MFCD00046592
InChI Key YBQOTTAEVBHNEJ-UHFFFAOYSA-N
Synonym 2-phenyl-2-pentanol, 2-phenyl-pentan-2-ol, benzenemethanol, .alpha.-methyl-.alpha.-propyl, methyl phenylbutanol, acmc-1aei8, methyl phenyl n-propyl carbinol
PubChem CID 138214
IUPAC Name 2-phenylpentan-2-ol
SMILES CCCC(C)(C1=CC=CC=C1)O
Specifications
Quantity 5 g
Formula Weight 164.25
Percent Purity 98%
Chemical Name or Material 2-Phenyl-2-pentanol