Learn More
2,6-Dimethylcyclohexanone, cis + trans, 99%
Product Code. p-7020624
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
CAS: 2816-57-1 | C8H14O | 126.199 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2816-57-1
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.199
MDL Number MFCD00001637
InChI Key AILVYPLQKCQNJC-UHFFFAOYSA-N
Synonym 2,6-dimethylcyclohexanone, cyclohexanone, 2,6-dimethyl, cyclohexanone,6-dimethyl, 2,6-dimethyl cyclohexanone, 2,6-dimethyl-cyclohexanone, cyclohexanone,2,6-dimethyl, 2,6-dimethylcyclohexanone,c&t, 2,6-dimethyl-cyclohexan-1-one, 2,6-dimethylcyclohexanone cis + trans, 2,6-dimethylcyclohexanone, cis + trans
PubChem CID 17780
IUPAC Name 2,6-dimethylcyclohexan-1-one
SMILES CC1CCCC(C1=O)C
Specifications
Density 0.918
Boiling Point 174°C to 176°C
Flash Point 51°C (123°F)
Refractive Index 1.447
Quantity 25 g
UN Number UN1224
Beilstein 1099000
Formula Weight 126.2
Percent Purity 99%
Chemical Name or Material 2,6-Dimethylcyclohexanone, cis + trans